Learn More
Sartorius Octet AMC2 Biosensors / Case
Supplier:  Sartorius 185165
Encompass
Octet AMC2 Biosensors. Trays of 96 biosensors coated with Thermo ScientificTM CaptureSelectTM Biotin-Anti-AAVX conjugate for quantitation of AAV capsids. 5-tray pack includes a 10% per tray discount; 20-tray case includes a 20% per tray discount
Catalog No. 50-289-1709
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.