Learn More
Sartorius Octet AMC2 Biosensors / Tray
Supplier:  Sartorius 185163
Encompass
Octet AMC2 Biosensors. Trays of 96 biosensors coated with Thermo ScientificTM CaptureSelectTM Biotin-Anti-AAVX conjugate for quantitation of AAV capsids. 5-tray pack includes a 10% per tray discount; 20-tray case includes a 20% per tray discount
Catalog No. 50-289-1578