Learn More
Sartorius Octet Front Reagent Rack Seal
Supplier:  Sartorius 190108
Encompass
Octet Front Reagent Rack Seal. Silicon-based reusable septa for the Octet SF3 Front Reagent Rack. Designed to prevent any sample or reagent evaporation during long SPR assay run times.
Catalog No. 50-289-1753