Learn More
Sartorius Octet GST Biosensors / Case
Supplier:  Sartorius 185098
Encompass
The Octet Anti-Glutathione-S-Transferase GST Biosensors are coated with a high affinity anti-GST antibody and offer a rapid and label-free method for both quantitation of GST-tagged biomolecules and kinetic analysis of GST fusions and interaction partners. The high specificity of the antibody-based biosensor enables the direct quantitation of GST-tagged analytes in crude lysates column eluents cell lysates and cell culture supernatants serving as an alternative to traditional time-consuming analytical methods.
Catalog No. 50-289-1705
Add to Cart
Request a Quote