Learn More
Sartorius OCTET-R8 8-CHANNEL SYSTEM
Catalog No. 502891630 Shop All Sartorius Products
Encompass
The Octet R8 system is a versatile tool that can be used for a wide range of analyses such as kinetic analysis titer of IgG and other proteins reagent qualification immunoassay development bioprocess development quality control crude antibody screening epitope binning/mapping ligand binding assays small molecule analysis elucidating cell signaling mechanisms and infectious disease monitoring. You can perform analysis using a single channel or up to eight channels giving you flexibility in sample throughput.High Throughput and Flexibility: The Octet R8 system has eight independent channels allowing for maximum speed sensitivity and flexibility. It can perform analyses of up to 96 samples in as little as 30 minutes to 2.5 hours catering to different sample throughput needs. It's suitable for a range of analyses including kinetic analysis titer determination immunoassay development and more.
Catalog No. 50-289-1630 Supplier Sartorius Supplier No. OCTETR8
To request a quote for this item, please select Request a Quote to reach our dedicated support team.
The Octet R8 system is a versatile tool that can be used for a wide range of analyses such as kinetic analysis titer of IgG and other proteins reagent qualification immunoassay development bioprocess development quality control crude antibody screening epitope binning/mapping ligand binding assays small molecule analysis elucidating cell signaling mechanisms and infectious disease monitoring. You can perform analysis using a single channel or up to eight channels giving you flexibility in sample throughput.High Throughput and Flexibility: The Octet R8 system has eight independent channels allowing for maximum speed sensitivity and flexibility. It can perform analyses of up to 96 samples in as little as 30 minutes to 2.5 hours catering to different sample throughput needs. It's suitable for a range of analyses including kinetic analysis titer determination immunoassay development and more.