Learn More
Sartorius Octet Rear Reagent Rack Seal
Supplier:  Sartorius 190107
Encompass
Octet Rear Reagent Rack Seal. Silicon-based reusable septa for the Octet SF3 Front Reagent Rack. Designed to prevent any sample or reagent evaporation during long SPR assay run times.
Catalog No. 50-289-1665