Learn More
Sartorius The Octet® Regeneration Buffer Kit provides a set of four ready-to-use buffers for screening regeneration conditions for various Octet® Biosensors.
Catalog No. NC2957642 Shop All Sartorius Products
Encompass
The kit includes:10 mM Glycine, pH 1.0, 15 mL10 mM Glycine, pH 1.7, 15 mL40 mM Phosphoric Acid, 15 mL
Catalog No. NC2957642 Supplier Sartorius Supplier No. 185171
The kit includes:10 mM Glycine, pH 1.0, 15 mL10 mM Glycine, pH 1.7, 15 mL40 mM Phosphoric Acid, 15 mL