Learn More
Sartorius Octet SPR CDH Sensor Chip
Catalog No. 502891710 Shop All Sartorius Products
Encompass
Octet SPR sensor chips have been specifically designed to allow generation of robust and reproducible results for every application – from large molecules molecular complexes viruses or whole cells to small molecules or antibody fragments. Octet SPR CDH sensor chips can be used for almost any Octet SPR assay and feature target interactions with low binding activity. They are biocompatible with a wide range of molecules.
Catalog No. 50-289-1710 Supplier Sartorius Supplier No. 190128
Octet SPR sensor chips have been specifically designed to allow generation of robust and reproducible results for every application – from large molecules molecular complexes viruses or whole cells to small molecules or antibody fragments. Octet SPR CDH sensor chips can be used for almost any Octet SPR assay and feature target interactions with low binding activity. They are biocompatible with a wide range of molecules.