Learn More
Sartorius Octet SPR Clean Station Cover
Catalog No. 502891628 Shop All Sartorius Products
Encompass
Octet SPR Clean Station Cover. Replacement cover for the Octet SF3 wash station to ensure injection needle is kept clean at all times.
Catalog No. 50-289-1628 Supplier Sartorius Supplier No. 190106
Octet SPR Clean Station Cover. Replacement cover for the Octet SF3 wash station to ensure injection needle is kept clean at all times.