Learn More
Sartorius Octet SPR COOH1 Sensor Chip
Supplier: Sartorius 190053
Octet SPR sensor chips have been specifically designed to allow generation of robust and reproducible results for every application – from large molecules molecular complexes viruses or whole cells to small molecules or antibody fragments. The Octet SPR COOH1 sensor chip can be used for whole cell virus particle and multivalent or large analyte assays. Its matrix-free surface makes it optimal for large molecular weight analytes.