Learn More
Sartorius Octet SPR HisCap Sensor Chip
Supplier:  Sartorius 190126
Encompass
Octet SPR sensor chips have been specifically designed to allow generation of robust and reproducible results for every application – from large molecules molecular complexes viruses or whole cells to small molecules or antibody fragments.Octet SPR HisCap sensor chips are ideal for small molecule peptide or fragment kinetics and large molecule kinetic assays. They provide a reliable alternative for proteins unamenable to amine coupling.
Catalog No. 50-289-1664