Learn More
Sartorius Octet SPR PCH Sensor Chip
Supplier:  Sartorius 190129
Encompass
Octet SPR sensor chips have been specifically designed to allow generation of robust and reproducible results for every application – from large molecules molecular complexes viruses or whole cells to small molecules or antibody fragments. Octet SPR PCH sensor chips are useful for small molecules due to high signal to noise ratio and can be used in experimental conditions unfavorable to dextran polycarbonate surfaces.
Catalog No. 50-289-1725