Learn More
Sartorius Octet SPR Plate Seal
Catalog No. 502891684 Shop All Sartorius Products
Encompass
Octet SPR Plate Seal Pack of 5 self-adhesive transparent microfoils compatible with polystrene and polypropylene plates and are designed to prevent any sample or reagent evaporation during long SPR assay run times.
Catalog No. 50-289-1684 Supplier Sartorius Supplier No. 190104
Octet SPR Plate Seal Pack of 5 self-adhesive transparent microfoils compatible with polystrene and polypropylene plates and are designed to prevent any sample or reagent evaporation during long SPR assay run times.