Learn More
Sartorius Octet SPR Vial 7.5mL
Catalog No. 502891682 Shop All Sartorius Products
Encompass
Octet SPR Glass Vial 7.5 mL. Manufactured using heat and chemical resistant borosilicate glass Octet SPR vials help you reduce your need for single-use plastics. With small dead volumes they help minimise the need to prepare excess sample. 7.5 mL vials are ideal for commonly used reagents in assays such as buffer or regeneration solution injections and can be used in conjunction with Octet SF3 Sample & Reagent Racks where sample evaporation is minimized via a reusable septa.
Catalog No. 50-289-1682 Supplier Sartorius Supplier No. 190143
Octet SPR Glass Vial 7.5 mL. Manufactured using heat and chemical resistant borosilicate glass Octet SPR vials help you reduce your need for single-use plastics. With small dead volumes they help minimise the need to prepare excess sample. 7.5 mL vials are ideal for commonly used reagents in assays such as buffer or regeneration solution injections and can be used in conjunction with Octet SF3 Sample & Reagent Racks where sample evaporation is minimized via a reusable septa.