Learn More
Sartorius Octet SPR Vial 7.5mL Set of 3
Catalog No. 502891615 Shop All Sartorius Products
Encompass
Octet SPR Glass Vial 7.5 mL Set of 3. Manufactured using heat and chemical resistant borosilicate glass Octet SPR vials help you reduce your need for single-use plastics. With small dead volumes they help minimise the need to prepare excess sample. 7.5 mL vials are ideal for commonly used reagents in assays such as buffer or regeneration solution injections and can be used in conjunction with Octet SF3 Sample & Reagent Racks where sample evaporation is minimized via a reusable septa.
Catalog No. 50-289-1615 Supplier Sartorius Supplier No. 190142
Octet SPR Glass Vial 7.5 mL Set of 3. Manufactured using heat and chemical resistant borosilicate glass Octet SPR vials help you reduce your need for single-use plastics. With small dead volumes they help minimise the need to prepare excess sample. 7.5 mL vials are ideal for commonly used reagents in assays such as buffer or regeneration solution injections and can be used in conjunction with Octet SF3 Sample & Reagent Racks where sample evaporation is minimized via a reusable septa.