Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octisalate, USP, 95-105%, Spectrum™ Chemical
SDP

Catalog No. 18604871 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
4 L
20 kg
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-871 4 L Amber Glass Bottle
18-604-870 500 mL Amber Glass Bottle
18-604-872 20 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-604-871 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. O12364LT
Add to Cart
Add to Cart

O1236, 118-60-5, C15H22O3

Spectrum™ Chemical Octisalate, USP absorbs ultraviolet radiation in the UVB range and is used in sunscreens and other cosmetics to protect from skin damage. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 118-60-5
Molecular Formula C15H22O3
Molecular Weight (g/mol) 250.34
InChI Key FMRHJJZUHUTGKE-UHFFFAOYNA-N
IUPAC Name 2-ethylhexyl 2-hydroxybenzoate
SMILES CCCCC(CC)COC(=O)C1=CC=CC=C1O

Specifications

Acidity 0.2 mL
CAS Max % 1
Packaging Amber Glass Bottle
Refractive Index 1.500-1.503
Quantity 4 L
Specific Gravity 1.011-1.016
Percent Purity 95 to 105%
Grade USP
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.