Learn More
CPC Scientific H-DPhe-Cys-Phe-DTrp-Lys-Thr-Cys-Thr-ol 1MG
SDP
Supplier:  CPC Scientific SOMA004B
Encompass
The synthetic analog of somatostatin that is three times more potent than the native hormone in inhibiting the secretion of growth hormone.SEQUENCE: H-DPhe-Cys-Phe-DTrp-Lys-Thr-Cys-Thr-ol (Disulfide Bridge)ONE-LETTER SEQUENCE: fCFwKTCT-ol (C2&C7 bridge)MOLECULAR FORMULA: C48H66N10O9S2MOLECULAR WEIGHT: 1019.26STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [79517-01-4]SYNONYMS: Octreotide, SMS 201-995, (D-Phe5,Cys6,11,D-Trp8,L-Threoninol12)-Somatostatin-14 (5-12)RESEARCH AREA: GastrointestinalREFERENCES:S.Grozinsky-Glasberg et al., Neuroendocrinology, 87, 168 (2008)T.E.Jonassen et al., Am. J. Physiol. Renal Physiol., 291, F537 (2006)H.G.Ding et al., Chin. Med. J., 117, 913 (2004)G.Melacini et al., Biochemistry, 36, 1233 (1997)S.W.Lamberts et al., N. Engl. J. Med., 334, 246 (1996)D. Maouyo et al., Pancreas, 14, 47 (1997)W. Bauer et al., Life Sci, 31(11), 1133 (1982)SKU(s): SOMA-004A, SOMA-004B
Catalog No. 50-211-0426
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.