Learn More
Sigma Organic Chemistry Octyl acrylate | 100mg | 2499-59-4 | MFCD00048935
Supplier: Sigma Organic Chemistry CDS004485100MG

Octyl acrylate , 100mg
About this Item:
CAS #: 2499-59-4
MDL #: MFCD00048935
Molecular Weight: 184.28
UNSPSC Code: 12352200
Chemical Formula: C11H20O2
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.