Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Octyltrichlorosilane, 97%

Catalog No. AC216500050
Change view
Click to view available options
Quantity:
5 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC216500050 5 g
AC216501000 100 g
AC216505000 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC216500050 Supplier Thermo Scientific Chemicals Supplier No. 216500050

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

CAS: 5283-66-9 | C8H17Cl3Si | 247.67 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5283-66-9
MDL Number MFCD00000488
InChI Key RCHUVCPBWWSUMC-UHFFFAOYSA-N
Synonym trichloro octyl silane, n-octyltrichlorosilane, octyltrichlorosilane, silane, trichlorooctyl, trichloro-n-octylsilane, unii-jv18i1wcu8, jv18i1wcu8, silane, octyltrichloro, octyltrichlorosilane un1801 corrosive
PubChem CID 21354
IUPAC Name trichloro(octyl)silane
SMILES CCCCCCCC[Si](Cl)(Cl)Cl

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 1.07g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C8H17Cl3Si
Refractive Index 1.4470 to 1.4490 (20°C, 589nm)
Linear Formula CH3(CH2)7SiCl3
Quantity 5 g
Beilstein 04, III, 1907
Molecular Weight (g/mol) 247.659
Formula Weight 247.67
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material Octyltrichlorosilane
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS P Statement
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.