Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ODC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA521362 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA521362 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA521362 Supplier Invitrogen™ Supplier No. PA521362
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: mouse brain, rat brain. Predicted reactivity: Mouse (91%), Rat (91%), Xenopus laevis (81%), Pig (94%), Chicken (85%), Rhesus Monkey (98%), Chimpanzee (100%), Bovine (93%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Ornithine decarboxylase catalyzes the conversion of ornithine to putrescine in the first and apparently rate-limiting step in polyamine biosynthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ODC1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.19 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene Odc1
Gene Accession No. P00860, P09057, P11926
Gene Alias MGC103389; Odc; ODC1; Ornithine decarboxylase; ornithine decarboxylase 1; ornithine decarboxylase, structural 1; Ornitine decarboxylase
Gene Symbols Odc1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human ODC. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18263, 24609, 4953
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.