Learn More
Description
- Can be used as ready-to-use spray or diluted for large area use
- Neutralizes strong odors
- Does not contains harmful fluorocarbons
Specifications
Specifications
| Form | Concentrated Liquid |
| Description | OdorAider deodorant |
| For Use With (Application) | Neutralizes strong odor |
| Volume (English) | 32 oz. |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.