Learn More
Enzo Life Sciences ODQ (10mg). CAS: 41443-28-1
SDP
Supplier:  Enzo Life Sciences ALX270034M010
Encompass_Preferred
ODQ is a potent and selective, direct-acting inhibitor of guanylate cyclase (IC50=20 nM). In cerebellar slices ODQ reversibly inhibited cGMP generation in response to endogenous NO or exogenously added NO-donors. ODQ does not inhibit NO-mediated macrophage toxicity (a phenomenon unrelated to cGMP) and does not inhibit particulate guanylyl cyclase or adenylyl cyclase. ODQ is an unique and selective tool to elucidate the physiological importance of the NO-cGMP pathway. Formula: C9H5N3O2. MW: 187.2. Purity: ≥98% (TLC). Solubility: Soluble in DMSO (5mg/ml), ethyl acetate or 100% ethanol (1.2mg/ml); insoluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-8373
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.