Learn More
Sellstrom™ Odyssey II Cleanroom Goggles
Catalog No. 19823050
Click to view available options
Ventilation Details:
Non-vent
Non-vent body
Lens Tint:
Clear
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Ventilation Details Lens Tint
19-823-050 Non-vent body Clear
19-822-925 Non-vent Clear
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
†Autoclaving is NOT recommended for goggles with anti-fog lenses (Cat. No. 19-822-925 and 19-823-051).
Catalog No. 19-823-050 Supplier Sellstrom™ Supplier No. S80231
This item is not returnable. View return policy
†Autoclaving is NOT recommended for goggles with anti-fog lenses (Cat. No. 19-822-925 and 19-823-051).
Sterilize by autoclaving with steam or gamma radiation†
  • For aseptic and cleanroom conditions; designed to be easily cleaned
  • Great panoramic view
  • High temperature, nonvented and top-vented body options available
  • UV-absorbing polycarbonate, durable antifog coating
  • Ultra comfortable, soft goggle body for a snug seal
  • Easily fits over prescription eyewear
  • Made of anallergic materials
  • Easy strap adjustment
Order Info
Replacement parts available (See Cat. No. 19-823-051).
Specifications
Lens Coating Uncoated
Lens Tint Clear
Product Type Cleanroom Goggles
Frame Color Blue
Ventilation Details Non-vent body
Material Polycarbonate
Frame Material TPR