Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ OF 1

Catalog No. 528950
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-895-0 50 mg
52-891-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 52-895-0 Supplier Tocris Bioscience™ Supplier No. 5289/50
This item is not returnable. View return policy

Selective BRPF1B and BRPF2 inhibitor

OF 1 is a selective BRPF1B and BRPF2 bromodomain inhibitor (Kd values are 100 and 500 nM respectively). Exhibits 39-fold selectivity for BRPF1B and BRPF2 over BRD4. Accelerates FRAP recovery at 5 μM in a BRPF2 FRAP assay. Inhibits RANKL-induced differentiation of primary murine bone marrow cells and human primary monocytes into osteoclasts.

Specifications

Purity 98%
Molecular Formula 4-Bromo-N-(2,3-dihydro-6-methoxy-1,3-dimethyl-2-oxo-1 H-benzimidazol-5-yl)-2-methylbenzenesulfonamide
CAS 919973-83-4
Content And Storage 4°C
Quantity 50 mg
Product Type OF 1
Molecular Weight (g/mol) M.W. (Observed): 440.31
Inhibitors OF 1

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.