Learn More
FUJIFILM IRVINE SCIENTIFIC INC iCell GlutaNeurons, 01279, 1 Vial: > 6 x 106 cells/vial
Supplier:  FUJIFILM IRVINE SCIENTIFIC INC R1034
Encompass
iCell GlutaNeurons, human glutamatergic-enriched cortical neurons derived from induced pluripotent stem (iPS) cells, display typical physiological characteristics and form functional neuronal networks amenable to examination across a number of commonly used assay techniques.
Catalog No. NC1400790
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.