Learn More
TARGETMOL CHEMICALS INC OGERIN 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T16378
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Ogerin is a selective GPR68 positive allosteric modulator (pEC50 6.83). Ogerin blocks recall in fear conditioning in mice. Ogerin displays inverse agonist and antagonist activity (Ki 220 nM) at A2A receptor and weak antagonist activity (Ki 736 nM) at 5-HT2B receptor. purity: 98%
Catalog No. 50-257-7340
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.