Learn More
Ohio Valley Specialty Chemical Gas Chromatography Septa
Catalog No. 22289343
Change view
Click to view available options
Type:
Septa
Diameter (Metric) Inner:
11 mm
Catalog No. Type Diameter (Metric) Inner
22-289343 Septa 11 mm
1 options
Catalog No. 22-289343 Supplier Ohio Valley Specialty Chemical Supplier No. 237715
Add to Cart
Add to Cart
Molded for dimensional consistency, giving more injections per septum
  • Low bleed, high temperature septa
  • Soft and resealable
Specifications
Type Septa
Diameter (Metric) Inner 11 mm