Learn More
Showa Denko America Inc OHpak LB-806, SEC chromatography column, 8.0 x 300 mm, Polyhydroxymethacrylate gel, 13 μm particle size, Pressure 3.5 MPa, Flow rate 0.5 to 1.2 mL/min, Temperature range 4 to 80 °C, pH 3 to 10, Max MW range 100,000 - 20,000,000
Supplier:  Showa Denko America Inc F6429205
Encompass_Preferred
The OHpak LB-806 is a high-performance, polymer-based packed column designed for aqueous Size Exclusion Chromatography (SEC) and Gel Filtration Chromatography (GFC). This robust column excels in applications utilizing light scattering detectors, thanks to its controlled low bleeding. It is compatible with versatile eluents, including N,N-Dimethylformamide (DMF) for polar polymers and meets USP-NF L38 and L39 requirements for regulated methods.
  • Aqueous SEC/GFC analysis for water-soluble macromolecules.
  • MALS compatibility with low column bleeding for sensitive detectors.
  • Can utilize DMF as an eluent for polar polymer analysis.
  • Fulfills USP-NF L38 and L39 requirements.
Catalog No. NC2888635