Learn More
New Pig Oil-Only Spill Kit in 95-Gallon Overpack Salvage Drum
SDP
Catalog No. 18389125 Shop All New Pig Corporation Products
Change view
Click to view available options
Type: Oil-Only Spill Kit in 95-Gallon Overpack Salvage Drum
Catalog No.
18-389-125 Type: Oil-Only Spill Kit in 95-Gallon Overpack Salvage Drum
1 options
This item is not returnable. View return policy
Catalog No. 18-389-125 Supplier New Pig Supplier No. KIT402
Add to Cart
Add to Cart
This item is not returnable. View return policy
Strong enough to earn a UN rating for shipping spill cleanup waste.
  • Comply with Spill Plan regulations, avoid fines and be ready to respond with the #1 kit for oil-based spill
  • Overpack drum container is UN rated for shipping waste after oil spill cleanup
  • Lift-out, prepacked baskets speed access and guard contents from UV
  • Absorbents meet ANSI and MIL-SPEC standards for static decay to allow use with fuels and flammables
  • Overpack is X-rated in Packing Groups I, II and III for shipping spill cleanup waste by land, sea or rail
  • Lightweight, polyethylene container resists chemicals and keeps contents clean and dry
  • Leakproof, twist-on lid is notched for easy removal or tightening with a 2x4 or pole
  • Bold lettering for quick identification
  • Ledges molded into container make kit easy to move by forklift
  • Tamperproof seals help prevent pilfering of spill response supplies
Specifications
Material Polyethylene