Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oil Red O, Sigma-Aldrich

Catalog No. 01258891 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-891 100 g
01-258-892 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-258-891 Supplier MilliporeSigma Supelco Supplier No. O0625100G
Add to Cart
Add to Cart

certified by the BSC

Oil Red O has been used for the staining of lipid droplets and lipid-containing vacuoles in cells.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1320-06-5
Molecular Weight (g/mol) 408.49 g/mol
Grade Certified
Color Red to Rust
Quantity 100 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.