Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oil Red O Solution, Sigma-Aldrich

Catalog No. 01258897 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
250 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-897 250 mL
01-258-973 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-258-897 Supplier MilliporeSigma Supelco Supplier No. O1391250ML
Add to Cart
Add to Cart

Oil red O solution has been used to stain neutral lipids and adipocytes and to visualize fat droplets in fibroblasts.

TRUSTED_SUSTAINABILITY

Specifications

CAS 1320-06-5
Concentration 0.5% in Isopropanol
Molecular Weight (g/mol) 408.49 g/mol
Color Dark Red
Quantity 250 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.