Learn More
Justrite™ Oily Waste Can With Hand-Operated Cover, Red
GSA_VA
Catalog No. 19822439 Shop All Justrite Safety Group Products
Change view
Click to view available options
Capacity (English):
10 gal.
14 gal.
Height (English):
18.3 in.
20.3 in.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (English) Height (English)
19-822-439 10 gal. 18.3 in.
19-822-440 14 gal. 20.3 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 19-822-439 Supplier Justrite™ Supplier No. 09310
Add to Cart
Add to Cart
Increase workplace safety and improve cleaniliness with the proper safety and storage equipment. The Justrite™ Oily Waste Can With Hand-Operated Cover, Red is an ideal solution for the disposal of oil-soaked rags. Prevent fires caused by stray sparks or spontaneous combustion.
  • Rags and cloths soaked with solvents, thinners, linseed oil, combustible adhesives and other flammable liquids present a serious fire risk when improperly discarded
  • Lid opens to no more than 60 degrees and stays closed when not in use, isolating contents from fire sources and limiting oxygen to virtually eliminate the risk of spontaneous combustion
  • Round construction and elevated base allow circulation of air around can to disperse heat and reduce moisture build-up and rusting
  • Durable steel construction with a powder-coat finish provides chemical resistance
  • Ten-year warranty.
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Capacity (English) 10 gal.
Color Red
Height (English) 18.3 in.