Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ OIP5 Monoclonal Antibody (OTI1D12)
GREENER_CHOICE

Catalog No. PIMA527335
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA527335 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA527335 Supplier Invitrogen™ Supplier No. MA527335
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

OIP5 Monoclonal Antibody for Western Blot, IHC (P)

Required for recruitment of CENPA to centromeres and normal chromosome segregation during mitosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen OIP5
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI1D12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Gene OIP5
Gene Accession No. O43482
Gene Alias 5730547N13Rik; cancer/testis antigen 86; CT86; hMIS18beta; LAP2alpha interactor-25; LINT-25; MIS18 kinetochore protein homolog B; MIS18B; MIS18beta; OIP5; OIP-5; Opa interacting protein 5; opa-interacting protein 5; Protein Mis18-beta; RGD1564263
Gene Symbols OIP5
Host Species Mouse
Immunogen Full length human recombinant protein of OIP5 produced in E.coli.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11339
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.