Learn More
OK-1™ Half-Finger Anti-Vibration Gloves
Description
- A precurved shape and half finger design provide dexterity
- Gloves offer superior protection from sources of vibration and impact
- Maintain a consistent grip due to the curved design that forms to the natural shape of the hand
- Machine washable
Specifications
Specifications
| Color | Brown |
| Hand | Specific |
| Material | Grain Cowhide Leather |
| Product Type | Work Gloves |
| Size | Large |
| For Use With (Application) | Construction, Machine Operation, and Manufacturing |
| Series | OK980 |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.