Learn More
Enzo Life Sciences Okadaic acid (high purity), (25µg), CAS Number: 78111-17-8
Supplier: Enzo Life Sciences ALX350003C025
Inhibitor of PP1 and PP2A. Alternative Name: Halochondrine A, 9,10-Deepithio-9,10-didehydroacanthifolicin. Formula: C44H68O13. MW: 805.0. CAS Number: 78111-17-8. MI: 14: 6819. Purity: ≥98% (TLC). Appearance: White crystalline solid. Solubility: Soluble in chloroform, ethanol, methanol, acetone or ethyl acetate. Use/Stability: Use only fresh solutions. Long Term Storage: -20°C
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.