Learn More
AdipoGen Okadaic acid Na high purity
SDP
Supplier:  AdipoGen AGCN20062C025
Encompass_Preferred
Chemical. CAS 209266-80-8. Formula C44H67O13 . Na. MW 804.0 . 23.0. Isolated from Prorocentrum concavum. Salt form generated in aqueous sodium hydroxide-methanol solution. Non-phorbol type tumor promoter. Reversible, potent and selective serine threonine protein phosphatase inhibitor. PP2A IC50=0.2-1nM, PP1 IC50=3-15nM, PP2B IC50=>1µM. Does not inhibit PP2C. Stimulates intracellular protein phosphorylation. Useful tool for studying cellular processes that are regulated by phosphorylation. Does not affect activity of acid phosphatase, alkaline phosphatase and tyrosine phosphatase. Mimics the effects of insulin. Activates atypical protein kinase C zeta/lambda in 3T3/L1 adipocytes. Enhances transmitter release at neuromuscular junctions. Apoptosis inhibitor. Induces apoptosis in human breast carcinoma cells MB-231 and MCF-7 and in myeloid cells. Neurotoxic.
Catalog No. 50-256-2676
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.