Learn More
Apexbio Technology LLC Olanzapine 132539-06-1 5g
Supplier: Apexbio Technology LLC B22405000
Olanzapine (CAS 132539-06-1) is a small molecule antagonist with high affinity for serotonin 5-HT2 and dopamine D2 receptors both classified as G protein-coupled receptors (GPCRs) In vitro binding studies demonstrate Olanzapine interacts at nanomolar concentrations with dopaminergic serotonergic 1-adrenergic and muscarinic receptors In vivo Olanzapine antagonizes dopamine and serotonin receptors and exhibits weaker efficacy at -adrenergic and muscarinic sites Administration in animal models increases extracellular dopamine and norepinephrine levels in specific brain regions in a dose-dependent manner Olanzapine is widely utilized in neuropharmacological research to investigate antipsychotic mechanisms
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.