Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Olanzapine, Atypical Antipsychotic, >99% Purity, Solid, 10 mg

Catalog No. 502707322
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7322 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7322 Supplier Abcam Supplier No. AB12073610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Atypical antipsychotic with anxiolytic activity and >99% purity.

  • Molecular Weight: 312.43 g/mol
  • Purity: >99%
  • Form: Solid
  • Biological Activity: 5-HT and D2 antagonist; displays activity at M1, M2, M3, M4, H1, α1 and α2 receptors
  • Ki Values (nM): 5-HT2A (4), 5-HT2C (11), 5-HT3 (57), D1 (31), D2 (11), D4 (27), M1 (1.9), M2 (18), M3 (25), M4 (13), α1 (19), α2 (7)
  • Classification: Atypical antipsychotic; displays anxiolytic activity

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Buffer 100% Olanzapine
Content And Storage Store at -20°C.
Form Solid
Shipping Condition Ambient
Target Molecule 5-HT Receptor, Dopamine Receptor, Histamine Receptor, Muscarinic Acetylcholine Receptor
Formulation 100% Olanzapine
Quantity 10 mg
For Use With (Application) Immunocytochemistry, Immunofluorescence
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.