Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Olanzapine, Atypical Antipsychotic, Solid, >99% Purity, 50 mg

Catalog No. 502707321
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-7321 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-7321 Supplier Abcam Supplier No. AB12073650MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Atypical antipsychotic compound with anxiolytic activity and >99% purity.

  • MW: 312.43 g/mol
  • Purity: >99%
  • Form: Solid
  • Activity: Atypical antipsychotic and anxiolytic
  • Antagonist Activity: 5-HT and D2 antagonist (Ki values: 4 (5-HT2A), 11 (5-HT2C), 57 (5-HT3), 31 (D1), 11 (D2), 27 nM (D4))
  • Receptor Activity: M1, M2, M3, M4, H1, α1 and α2 receptors (Ki values: 1.9 (M1), 18 (M2), 25 (M3), 13 (M4), 19 (α1), 7 nM (α2))
  • Storage: -20°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule 5-HT Receptor, Dopamine Receptor, Histamine Receptor, Muscarinic Acetylcholine Receptor, α-Adrenergic Receptor
Shipping Condition Ambient
Substrate Properties Chemical Substrate
Content And Storage Store at -20°C.
Form Solid
Quantity 50 mg
For Use With (Application) Antipsychotic, Anxiolytic
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.