Learn More
Cayman Chemical Oleic Acid, 112-80-1, 1 g
SDP
Supplier:  Cayman Chemical 902601
Encompass_Preferred
Molecular Formula C18H34O2, Purity ≥98%, Formula Weight 282.5, Synonyms: ​cis-9-Octadecenoic acid; A monounsaturated fatty acid; a major component of membrane phospholipids; inhibits collagen-stimulated platelet aggregation at 10 g/ml; inhibits fMLF-induced neutrophil aggregation and degranulation at 5 M; induces release of intracellular calcium in human platelets at 60 M; increases TNF-, IL-8, IL-6, and IL-1 production, neutrophil accumulation, and apoptotic and necrotic cell death in mouse lung.
Catalog No. NC0682989
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.