Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oleic Acid, Purified, (cis-9-Octadecenoic Acid), Reagents

Catalog No. 01337581
Change view
Click to view available options
Quantity:
100 mL
500 mL
1 L
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-581 500 mL
01-337-583 100 mL
01-337-582 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 01-337-581 Supplier Reagents Holdings Llc Supplier No. C2110000500C
Add to Cart
Add to Cart

Oleic Acid, Purified, (cis-9-Octadecenoic Acid), Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material Oleic Acid, Purified, (cis-9-Octadecenoic Acid)
CAS 112-80-1
Boiling Range 286°C
Melting Point 14°C
Color Colorless to Slight Yellow
Boiling Point 286°C
Melting Range 14°C
Packaging Glass Bottle
Molecular Formula C18H34O2
Quantity 500 mL
Specific Gravity 0.895
Synonym cis-9-Octadecenoic Acid
Molecular Weight (g/mol) 282.2559
Grade Reagent
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.