Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oleyl Alcohol, Approx. 99%, MP Biomedicals™

Catalog No. ICN15599690 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15599690 500 mg
ICN15599691 1 g
ICN15599694 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15599690 Supplier MP Biomedicals, Inc Supplier No. 0215599690
Add to Cart
Add to Cart

Chemical Identifiers

CAS 143-28-2
Molecular Formula C18H36O
Molecular Weight (g/mol) 268.49
MDL Number MFCD00002993
InChI Key ALSTYHKOOCGGFT-KTKRTIGZSA-N
Synonym oleyl alcohol, cis-9-octadecen-1-ol, z-octadec-9-en-1-ol, ocenol, conditioner 1, dermaffine, lancol, novol, oceol, oleol
PubChem CID 5284499
ChEBI CHEBI:73504
IUPAC Name (9Z)-octadec-9-en-1-ol
SMILES CCCCCCCC\C=C/CCCCCCCCO

Specifications

Color Colorless
Density 0.85g/mL
Assay Percent Range Approx. 99%
Quantity 500 mg
Formula Weight 268.49
Physical Form Liquid
Chemical Name or Material Oleyl Alcohol
EINECSNumber 205-597-3
Recommended Storage Store at -20°C.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.