Learn More
Sigma Organic Chemistry Oleylamine | 100G | 112-90-3 | MFCD00066507 | 0.7
Supplier: Sigma Organic Chemistry O7805100G

Oleylamine, 100G
About This Item:
Synonym(s): cis-1-Amino-9-octadecene
Linear Formula: CH3(CH2)7CH=CH(CH2)7CH2NH2
Molecular Weight: 267.49
CAS Number: 112-90-3
MDL Number: MFCD00066507
UNSPSC Code: 12162002
Purity: 0.7
Boiling Point: 348-350 C (lit.)
Melting Point: 18-26 C (lit.)
Density: 0.813 g/mL at 25 C (lit.)
Refractive Index: n20/D 1.4596 (lit.)
Storage: room temp
EINECS Number: 204-015-5
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.