Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ Oleylethanolamide
GREENER_CHOICE

Catalog No. 148410
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
14-841-0 10 mg
14-845-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 14-841-0 Supplier Tocris Bioscience™ Supplier No. 1484/10
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Lipid mediator, anorexic actions; also GPR55 and PPARα agonist

Lipid mediator and analog of anandamide (Cat. No. 1339) that is involved in peripheral regulation of feeding. Selective GPR55 agonist (EC50 values are 0.44, >30 and >30 μM at GPR55, CB1 and CB2 respectively) and PPARα agonist (EC50 = 120 nM). Induces satiety through activation of PPARα and is also a ceramidase inhibitor. Also endogenous agonist at the GPR119 receptor.

Chemical Identifiers

CAS 111-58-0
Molecular Formula C20H39NO2

Specifications

Quantity 10 mg
Formula Weight 325.53
Purity >99%
Molecular Weight (g/mol) 325.537
Chemical Name or Material Oleylethanolamide
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.