Learn More
Sigma Aldrich Fine Chemicals Biosciences Oli-neu Mouse Oligodendroglial Precursor Cell Line | CAS N/A | MDL N/A | 1 EA
Supplier:  Sigma Aldrich Fine Chemicals Biosciences SCC261
Encompass
Oli-neu mouse oligodendroglial precursor cell line | Purity: N/A | Mol Wt: N/A | CAS No: N/A | MDL No: N/A | N/AImmortalized mouse oligodendroglial precursor cell line that recapitulates oligodendroglial morphology and physiology. Differentiates into mature oligodendrocytes and expresses myelin proteins upon addition of dibutyrl-cAMP. Provides in vitro model system for studying oligodendrocyte biology and myelination.
Catalog No. NC2471913