Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Oligodendrocyte Marker O1 Monoclonal Antibody (O1), eFluor™ 660, eBioscience™, Invitrogen™

Catalog No. p-7092307
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-112-9284 25 μg
50-112-9062 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-112-9284 Supplier Invitrogen™ Supplier No. 50650680
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

Description: This O1 monoclonal antibody reacts with galactocerebroside and other lipids found on late oligodendrocyte precursors. O1 has been reported to react to all species tested (rat, mouse, human, chicken). Oligodendrocytes function as myelin-producing cells within the central nervous system ensheathing the axons of neurons with myelin in order to facilitate rapid axonal conduction. O1 and O4 monoclonal antibodies serve as stage-specific markers of oligodendrocyte development with O4 marking early oligodendrocyte progenitors and O1-labeling late oligodendrocyte progenitors. O4-positive cells differentiate to become O1-positive, which can in turn become mature oligodendrocytes. Applications Reported: This O1 antibody has been reported for use in flow cytometric analysis, immunohistochemical staining of frozen tissue sections, and immunocytochemistry. Applications Tested: This O1 antibody has been tested by immunocytochemistry on fixed, differentiated OLN93 cells and can be used at less than or equal to 20 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. eFluor™ 660 is a replacement for Alexa Fluor™ 647. eFluor™ 660 emits at 659 nm and is excited with the red laser (633 nm). Please make sure that your instrument is capable of detecting this fluorochome. Excitation: 633-647 nm; Emission: 668 nm; Laser: Red Laser. Filtration: 0.2 μm post-manufacturing filtered.

Oligodendrocytes function as myelin-producing cells within the central nervous system ensheathing the axons of neurons with myelin in order to facilitate rapid axonal conduction. O1 and O4 monoclonal antibodies serve as stage-specific markers of oligodendrocyte development with O4 marking early oligodendrocyte progenitors and O1-labeling late oligodendrocyte progenitors. O4-positive cells differentiate to become O1-positive, which can in turn become mature oligodendrocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Oligodendrocyte Marker O1
Applications Flow Cytometry, Immunohistochemistry (Frozen), Immunocytochemistry
Classification Monoclonal
Clone O1
Concentration 0.2 mg/mL
Conjugate eFluor 660
Formulation PBS with 0.09% sodium azide; pH 7.2
Gene Alias O1 Marker
Host Species Mouse
Purification Method Affinity chromatography
Quantity 25 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat, Chicken
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgM
Show More Show Less
Can the eFluor Organic fluorochromes be used for intracellular staining?

Yes, the eFluor Organic fluorochromes can be used for intracellular staining. The eFluor organic fluorochromes maintain bright signal and require minimal changes in compensation when fixed with eBioscience IC Fixation Buffer (Cat. No. 00-8222-49) and Permeabilization Buffer (Cat. No. 00-8333-56) or 1-step Fix/Lyse Solution (Cat. No. 00-5333-54, 00-5333-57) (as compared to live cells).

Is the eFluor 660 fluorochrome compatible with Anti-Cy5/Alexa Fluor 647 beads?

Yes, in-house studies have demonstrated that the eFluor 660 fluorochrome is recognized by Anti-Cy5/Alexa Fluor 647 beads. Side by side studies with Alexa Fluor 647 versus eFluor 660 conjugated antibodies have demonstrated comparable results.

What buffers are compatible with the eFluor Organic fluorochromes and eVolve QDots?

The eFluor Organic fluorochromes and eVolve QDots can be used with flow staining buffers containing PBS and protein.

Are the eFluor Organic Dyes photo-labile?

As with other fluorochromes, we recommend minimal exposure to light to maintain optimal signal.

For Research Use Only.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.