Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oligodendrocyte Marker O4 APC-conjugated Antibody, R&D Systems™
GREENER_CHOICE

Catalog No. FAB1326A
Change view
Click to view available options
Quantity:
100 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FAB1326A 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. FAB1326A Supplier R&D Systems Supplier No. FAB1326A
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Oligodendrocyte Marker O4 Monoclonal specifically detects Oligodendrocyte Marker O4 in Human, Mouse, Rat, Chicken samples. It is validated for Flow Cytometry.

Specifications

Antigen Oligodendrocyte Marker O4
Applications Flow Cytometry
Classification Monoclonal
Clone O4
Conjugate APC
Dilution Flow Cytometry 10 uL/10^6 cells
Formulation Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide
Host Species Mouse
Immunogen Bovine brain corpus callosum white matter
Purification Method Affinity Purified
Quantity 100 Tests
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Detects human, mouse, rat, and chicken Oligodendrocyte Marker O4.
Target Species Human, Mouse, Rat, Chicken
Content And Storage Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied.
Isotype IgM
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.