Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Oligodendrocytes Mouse anti-Rat, Clone: NS-1, MilliporeSigma™

Catalog No. MAB1580I100 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MAB1580I100 100 μL
MAB1580I25 25 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MAB1580I100 Supplier MilliporeSigma Supplier No. MAB1580I100UL
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Oligodendrocytes Monoclonal antibody specifically detects Oligodendrocytes in Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry, Immunofluorescence.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Oligodendrocytes
Applications Immunocytochemistry, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone NS-1
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation Purified mouse monoclonal antibody IgG in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Host Species Mouse
Immunogen Rat olfactory bulb cells.
Purification Method Protein G purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Rat
Content And Storage Stable for 1 year at +2°C to +8°C from date of receipt.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.