Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam Oligomycin A, ATP Synthase Inhibitor, >97% Purity, Solid, 5 mg

Catalog No. 502706117
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-6117 5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-6117 Supplier Abcam Supplier No. AB1434235MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Macrolide antibiotic and potent ATP synthase inhibitor that blocks the proton channel to uncouple oxidative phosphorylation.

  • Molecular Weight: 791.1 Da
  • Purity: >97%
  • Description: Macrolide antibiotic that acts as an ATP synthase inhibitor by blocking the proton channel (Fo subunit) to uncouple oxidative phosphorylation
  • Additional Biological Activity: Shows antitumor activity and is active in vivo
  • Form: Solid
  • Composition: 100% Oligomycin A

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Content And Storage Store at 4°C.
Shipping Condition Ambient
Target Molecule ATP Synthase
Form Solid
Formulation 100% Oligomycin A
Quantity 5 mg
For Use With (Application) Oxidative phosphorylation inhibition
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.