Learn More
Abcam Oligomycin A, ATP Synthase Inhibitor, Solid, 1 mg
Description
- Molecular Weight: 791.1 Da
- Purity: >97%
- Form: Solid
- Function: Macrolide antibiotic; acts as an ATP synthase inhibitor by blocking the proton channel (Fo subunit) to uncouple oxidative phosphorylation
- Additional Activity: Shows antitumor activity; active in vivo
- Storage: Store at +4°C
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Form | Solid |
| Target Molecule | ATP Synthase |
| Content And Storage | Store at 4°C. |
| Shipping Condition | Ambient |
| Formulation | 100% Oligomycin A |
| Quantity | 1 mg |
| For Use With (Application) | ATP synthase inhibition, oxidative phosphorylation uncoupling, antitumor research |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.